Cyclohexan for pesticide residue analysis
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Hexahydrobenzene, Hexamethylene
326590010 326590025
ACRO326590010EA 575 DKK
ACRO326590010 ACRO326590025
Cyclohexan for pesticide residue analysis
Cyclohexane
Formel: C₆H₁₂
Molekylvægt: 84.16 g/mol
Kogepunkt: 81
Smeltepunkt: 6.5 °C
Massefylde: 0.77 g/cm³
Flammepunkt: –18 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003814
CAS-nummer: 110-82-7
EINECS: 203-806-2
UN nummer: 1145
ADR: 3 II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM
Bestil nu
Specifikationer Testresultater
Appearance Clear colorless Liquid
Water ≤0.02 %
GC (ECD) Passes test, (NPD) Passes test, (FID) Passes test
Residue after evaporation ≤2 ppm