Cyclooctanol 97

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A17825.14 A17825.22
A17825.14EA 1220 DKK
A17825.14 A17825.22
Cyclooctanol 97
Cyclooctanol

Purity: 97% CAS No.: 696-71-9 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 86°(186°F) Refractive Index: 1.4860 Melting Point: 14-15° Boiling Point: 97-98°/11mm Beilstein Reference: 1848166 MDL No.: MFCD00001744

Formel: C₈H₁₆O
Molekylvægt: 128.21 g/mol
Kogepunkt: 97…98 °C (11 mmHg)
Smeltepunkt: 14…15 °C
Massefylde: 0.970
Flammepunkt: 86 °C (186 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001744
CAS-nummer: 696-71-9
EINECS: 211-800-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR