Cyclododecanol 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

110961000 110965000
ACRO110961000EA 485 DKK
ACRO110961000 ACRO110965000
Cyclododecanol 98+%
Cyclododecanol
Formel: C₁₂H₂₄O
Molekylvægt: 184.32 g/mol
Kogepunkt: 272.7
Smeltepunkt: 76…79 °C
Flammepunkt: 138 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003717
CAS-nummer: 1724-39-6
EINECS: 217-031-2
UN nummer: 3077
ADR: 9 III

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥98.0 %
Color scale ≤15 APHA
Titration with HCl ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR