Cyclopropylamin 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Aminocyclopropane
154880100
154880500
154882500
ACRO154880100EA
320
DKK
ACRO154880100
ACRO154880500
ACRO154882500
Cyclopropylamin 99%
Cyclopropylamin
Formel:
C₃H₇N Molekylvægt: 57.1 g/mol Kogepunkt: 49…50 °C (760 mmHg) Smeltepunkt: –50 °C Massefylde: 0.824 g/cm³ Flammepunkt: –25 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001301 CAS-nummer: 765-30-0 EINECS: 212-142-2 UN nummer: 2733 ADR: 3 II |
Specifikationer Testresultater
Appearance | Clear colorless Volatile liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4220 (20°C, 589 nm) |
Water | ≤0.5 % |
GC | ≥99 % |
Titration with HCl | 99.0 to 101.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...