1-Aminocyclopentancarboxylsyre 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
103470050
103470100
ACRO103470050EA
748
DKK
ACRO103470050
ACRO103470100
1-Aminocyclopentancarboxylsyre 97%
1-Aminocyclopentancarboxylsyre
Formel:
C₆H₁₁NO₂ Kogepunkt: 254 °C (1013 hPa) Smeltepunkt: 320…322 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001381 CAS-nummer: 52-52-8 EINECS: 200-144-6 UN nummer: 2811 ADR: 6.1,III Merck Index: 13,02763 |
Specifikationer Testresultater
Appearance | White to beige Crystalline powder or flakes |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
GC | ≥96.0 % |
Specific optical rotation | +42° to +48° (20°C, 589 nm) (c=3, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...