1,2-Cyclohexandion ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Purity: 98+%
CAS No.: 765-87-7
Molecular Formula: C6H8O2
Formula Weight: 112.13
Storage Temperature: 2°C to 8°C
Flash Point: 84°(183°F)
Refractive Index: 1.5190
Melting Point: 35-38°
Boiling Point: 193-195°
Beilstein Reference: 507419
MDL No.: MFCD00001648
Formel:
C₆H₈O₂ Kogepunkt: 193…195 °C Smeltepunkt: 35…38 °C Massefylde: 1,12 |
MDL Number:
MFCD00001648 CAS-nummer: 765-87-7 EINECS: 212-155-3 |