Cyclobutanon 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

110930010 110930050 110930250
110930010.EA 504 DKK
110930010. 110930050. 110930250.
Cyclobutanon 98+%
Cyclobutanone
Formel: C₄H₆O
Kogepunkt: 99 °C (760 mmHg)
Massefylde: 0,93 g/cm³
Flammepunkt: 10 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00001332
CAS-nummer: 1191-95-3
EINECS: 214-745-6
UN nummer: 1224
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4230 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR