7-Methylquinolin ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

42475.03 42475.01
42475.03EA 995 DKK
42475.03 42475.01
7-Methylquinolin ≥97%
7-Methylquinolin

Purity: 97% CAS No.: 612-60-2 Molecular Formula: C10H96N Formula Weight: 143.19 Flash Point: 110°(230°F) Physical Form: Viscous liquid Density: 1.07 Boiling Point: 251-253° Beilstein Reference: 110317 MDL No.: MFCD00006805

Formel: C₁₀H₉N
Molekylvægt: 143.19 g/mol
MDL Number: MFCD00006805
CAS-nummer: 612-60-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR