6-Methylquinolin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B21892.06 B21892.14 B21892.22
B21892.06EA 317 DKK
B21892.06 B21892.14 B21892.22
6-Methylquinolin ≥98%
6-Methylquinolin

Purity: 98% CAS No.: 91-62-3 Molecular Formula: C10H9N Formula Weight: 143.19 Flash Point: 110°(230°F) Refractive Index: 1.6150 Melting Point: -22° Boiling Point: 257-259° Beilstein Reference: 110336 MDL No.: MFCD00006804

Formel: C₁₀H₉N
Kogepunkt: 257…259 °C
Smeltepunkt: –22 °C
Massefylde: 1,066
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006804
CAS-nummer: 91-62-3
EINECS: 202-084-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR