Chinaldin 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Methylquinoline

A13412.14 A13412.22 A13412.36
A13412.14EA 354 DKK
A13412.14 A13412.22 A13412.36
Chinaldin 96%
Chinaldin

Used in preparation of oil-soluble dyes


It is an anaesthetic used in fish transportation. Also used in anti-malaria drugs, in manufacturing dyes, food colorants , pharmaceuticals and pH indicators.


Advarsel: Air sensitive. Light sensitive. Store under argon.

Formel: C₁₀H₉N
Molekylvægt: 143.19 g/mol
Kogepunkt: 247…248 °C
Smeltepunkt: –2 °C
Massefylde: 1.058
Flammepunkt: 79 °C (174 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006756
CAS-nummer: 91-63-4
EINECS: 202-085-1
Merck Index: 12,08228

Bestil nu

Specifikationer Testresultater

Appearance (Colour) Clear colorless to yellow to brown
Form Liquid
Assay (GC) ≥95,0%
Refractive Index 1,6080 - 1,6145 @ 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR