6-Fluorquinaldin ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A11469.06 A11469.14 A11469.22
A11469.06EA 1150 DKK
A11469.06 A11469.14 A11469.22
6-Fluorquinaldin ≥98%
6-Fluorquinaldin

Purity: 98+% CAS No.: 1128-61-6 Molecular Formula: C10H8FN Formula Weight: 161.18 Melting Point: 52-56° Beilstein Reference: 1281677

Formel: C₁₀H₈FN
Smeltepunkt: 52…56 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041233
CAS-nummer: 1128-61-6
EINECS: 214-439-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR