8-Aminoquinolin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Quinolin-8-ylamine
A16752.03
A16752.06
A16752.14
A16752.03EA
449
DKK
A16752.03
A16752.06
A16752.14
8-Aminoquinolin ≥98%
8-Aminoquinolin
Purity: 98+%
CAS No.: 578-66-5
Molecular Formula: C9H8N2
Formula Weight: 144.18
Melting Point: 63-67°
Boiling Point: 174°/26mm
MDL No.: MFCD00006809
Formel:
C₉H₈N₂ Molekylvægt: 144.18 g/mol Kogepunkt: 174 °C (26 mmHg) Smeltepunkt: 63…67 °C Massefylde: 1.337 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006809 CAS-nummer: 578-66-5 EINECS: 209-427-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...