3-Aminoquinolin 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Quinolin-3-amine, Quinolin-3-ylamine

213780050 213780010
ACRO213780050EA 695 DKK
ACRO213780050 ACRO213780010
3-Aminoquinolin 99%
3-Aminoquinolin
Formel: C₉H₈N₂
Kogepunkt: 311 °C (1013 hPa)
Smeltepunkt: 91…94 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006772
CAS-nummer: 580-17-6
EINECS: 209-455-1
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance White to beige to orange-brown Powder or crystals
Infrared spectrum Conforms
Melting point 92 to 96 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR