4,7-Dichlorquinolin ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A15985.14
A15985.22
A15985.36
A15985.14EA
806
DKK
A15985.14
A15985.22
A15985.36
4,7-Dichlorquinolin ≥98%
4,7-Dichlorquinolin
Purity: 98%
CAS No.: 86-98-6
Molecular Formula: C9H5Cl2N
Formula Weight: 198.05
Flash Point: 164°(327°F)
Melting Point: 83-87°
Boiling Point: 148°/10mm
Beilstein Reference: 125359
MDL No.: MFCD00006774
Formel:
C₉H₅Cl₂N Kogepunkt: 148 °C (10 mmHg) Smeltepunkt: 83…87 °C Flammepunkt: 164 °C (327 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00006774 CAS-nummer: 86-98-6 EINECS: 201-714-7 |