(1R)-(+)-α-Pinen ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 98%
CAS No.: 7785-70-8
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 32°(89°F)
Refractive Index: 1.4660
Melting Point: -62°
Boiling Point: 154-156°
Beilstein Reference: 3648264
Merck Reference: 14,7445
UN No.: 2368
MDL No.: MFCD00001346
Formel:
C₁₀H₁₆ Kogepunkt: 154…156 °C Smeltepunkt: –62 °C Massefylde: 0,858 Flammepunkt: 32 °C (89 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001346 CAS-nummer: 7785-70-8 EINECS: 232-087-8 UN nummer: 2368 ADR: 3,III Merck Index: 14,7445 |