Benzylbenzoat for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Benzoic acid benzyl ester
M-Clarity™ quality level = MQ200
8.18701.0100
8.18701.1000
8.18701.0100EA
190
DKK
8.18701.0100
8.18701.1000
Benzylbenzoat for synthesis, Sigma-Aldrich®
Benzylbenzoat
Formel:
C₁₄H₁₂O₂ Molekylvægt: 212.25 g/mol Kogepunkt: 323 Smeltepunkt: 17…20 °C Massefylde: 1.118 g/cm³ (20 °C) Flammepunkt: 158 °C (closed cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003075 CAS-nummer: 120-51-4 EINECS: 204-402-9 UN nummer: 3082 ADR: 9 III Merck Index: 13,01129 |
Specifikationer Testresultater
Solubility | 0.0153 g/l (20 °C) |
Melting Point | 21 °C |
Molar Mass | 212.24 g/mol |
Boiling Point | 324 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 158 °C |
Density | 1.12 g/cm3 (20 °C) |
Ignition Point | 480 °C |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...