4-Phenylcyclohexanon ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane

L08614.14 L08614.22
L08614.14EA 1090 DKK
L08614.14 L08614.22
4-Phenylcyclohexanon ≥98%
4-Phenylcyclohexanon

Purity: 98+% CAS No.: 4894-75-1 Molecular Formula: C12H14O Formula Weight: 174.24 Flash Point: 100°(212°F) Melting Point: 78-82° Boiling Point: 158-160°/12mm Beilstein Reference: 2045904 MDL No.: MFCD00001641

Formel: C₁₂H₁₄O
Kogepunkt: 158…160 °C (12 mmHg)
Smeltepunkt: 78…82 °C
Flammepunkt: >100 °C (212 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001641
CAS-nummer: 4894-75-1
EINECS: 225-517-0

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR