Saccharometers
Leverandør: GERING
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0700WG 0703SWG 0706WG 0712WG 0710WG 0701SWG
641-0000EA 0 DKK
641-0000 641-0002 641-0003 GERI0712WG 641-0004 641-0001
Saccharometers
Flydevægte
These saccharometers with temperature correction scale are suitable for the official testing in breweries especially on wort.