2,6-Di-tert-butyl-p-cresol ≥99%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Butylated hydroxytoluene, Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, BHT
A16863.0B A16863.30
A16863.0BEA 835 DKK
A16863.0B A16863.30
2,6-Di-tert-butyl-p-cresol ≥99%
2,6-Di-tert-butyl-p-cresol
Purity: 99% CAS No.: 128-37-0 Molecular Formula: C15H24O Formula Weight: 220.36 Flash Point: 127°(260°F) Odour: Mild Boiling Point: 264-265° Beilstein Reference: 1911640 Merck Reference: 14,1548 UN No.: 3077 MDL No.: MFCD00011644
Formel: C₁₅H₂₄O
Molekylvægt: 220.35 g/mol
Kogepunkt: 264…265 °C
Smeltepunkt: 69…72 °C
Massefylde: 1.048
Flammepunkt: 127 °C (260 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011644
CAS-nummer: 128-37-0
EINECS: 204-881-4
ADR: 9 III
Merck Index: 13,01547