Funnel chambers, CytoSep™, for Hettich Cyto-System
Leverandør: Simport Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M966-1 M966-8 M966-4 M966-2
720-1954EA 1020 DKK
720-1954 720-1873 720-1872 720-1955
Funnel chambers, CytoSep™, for Hettich Cyto-System
Tragte Cytologitragte
The CytoSep™ funnel chambers for the Hettich Cyto-System optimises laboratory throughput with multiple funnel options in 2, 3, or 4 funnels per slide.
  • These funnels are ideal for applications such as CSF, viral infections, or bronchial secretions, and are available in four sizes from 1 to 8 ml. The 1 ml chamber is for small volume samples of low cell content. Both 2 and 4 ml chambers are suggested for cell-rich samples, e.g. pleura, ascites and bronchial washings, while the 8 ml chamber is perfect for large volume samples such as urine.