Hand protectors, HotGrip
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9768-0001 9768-0002 9768-0003
BURK9768-0001EA 718 DKK
BURK9768-0001 BURK9768-0002 BURK9768-0003
Hand protectors, HotGrip
Håndbeskyttere
The cold and heat resistant HotGrip protectors are made of silicone rubber. They protect fingers and hands from burns and freezing when working with apparatus that have extreme temperatures from −60 to +200 °C (briefly up to +250 °C).
  • Safe handling and positioning of deep frozen, hot or even boiling substances and apparatus
  • Extremely elastic and slip-proof silicone rubber: Even smooth surfaced apparatus such as hot glass flasks can be securely gripped
The protectors are available for the whole hand, for thumb and one finger and for thumb and 1 to 2 fingers.