Aerosol-refiller til A1 luftfrisker, Tork Premium
Leverandør: Essity
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
236052 236050 236051 236056
115-1627EA 1710 DKK
115-1627 115-1625 115-1626 115-1628
Aerosol-refiller til A1 luftfrisker, Tork Premium
Deodoranter
Control odours effectively and efficiently in your washroom with the Tork Air Freshener Sprays. The sprays contain concentrated fragrance oils and proprietary odour neutralising agents to eliminate unpleasant smells while providing lasting air freshness.
The sprays are suitable for the Tork Air Freshener Spray Dispenser (A1).