Tork M1/M2 Roll Dispensers for Centrefeed
Leverandør: Essity
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
559008 558000 559000 558008 659000 659008 202040 200040
115-0607EA 791 DKK
115-0607 115-1679 115-0615 115-1680 115-1695 115-1696 115-1615 115-1614
Tork M1/M2 Roll Dispensers for Centrefeed
Holder til Servietter
These dispensers for centrefeed rolls enable an easy, one-handed operation while allowing a user to take as much paper as needed. They are ideal for areas where hand and light surface wiping are combined - such as in food preparation laboratories.
  • Easy load concept: Rapid refilling
  • Flexible dispensing: Take as much as you need