2,6-Diisopropylanilin 90+%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 90+%
CAS No.: 24544-04-5
Molecular Formula: C12H19N
Formula Weight: 177.29
Flash Point: 125°(257°F)
Refractive Index: 1.5330
Melting Point: -54°
Boiling Point: 254-257°
Beilstein Reference: 2208763
MDL No.: MFCD00008887
Formel:
C₁₂H₁₉N Kogepunkt: 254…257 °C Smeltepunkt: –54 °C Massefylde: 0,940 Flammepunkt: 125 °C (257 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008887 CAS-nummer: 24544-04-5 EINECS: 246-305-4 |