3-(Trifluormethyl)benzylchlorid ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 3-(Trifluoromethyl)benzyl chloride, 3-(Chloromethyl)benzotrifluoride, 1-(Chloromethyl)-3-(trifluoromethyl)benzene

A16253.06 A16253.14 A16253.22
A16253.06EA 411 DKK
A16253.06 A16253.14 A16253.22
3-(Trifluormethyl)benzylchlorid ≥98%
3-(Trifluormethyl)benzylchlorid

Purity: 98% CAS No.: 705-29-3 Molecular Formula: C8H6ClF3 Formula Weight: 194.59 Flash Point: 74°(165°F) Refractive Index: 1.4645 Odour: Pungent Boiling Point: 68-70°/12mm Beilstein Reference: 743242 UN No.: 3265 MDL No.: MFCD00000908

Formel: C₈H₆ClF₃
Molekylvægt: 194.58 g/mol
Kogepunkt: 68…70 °C (12 mmHg)
Massefylde: 1.254
Flammepunkt: 74 °C (165 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000908
CAS-nummer: 705-29-3
EINECS: 211-884-4
UN nummer: 3265
ADR: 8 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR