4-Methoxy-1-butanol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
4-Methoxybutan-1-ol
B2402-25G
B2402-5G
TCIAB2402-25GEA
1610
DKK
TCIAB2402-25G
TCIAB2402-5G
4-Methoxy-1-butanol ≥98.0% (by GC)
4-Methoxy-1-butanol
Formel:
C₅H₁₂O₂ Molekylvægt: 104.15 g/mol Kogepunkt: 66°C |
MDL Number:
MFCD00040437 CAS-nummer: 111-32-0 EINECS: 203-858-6 |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...