3-Aminobenzylamin ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
alpha-Amino-m-toluidine, 3-(Aminomethyl)aniline
A1431-25G
A1431.0005
A1431-5G
TCIAA1431-25GEA
731
DKK
TCIAA1431-25G
TCIAA1431.0005
TCIAA1431-5G
3-Aminobenzylamin ≥98.0% (by GC, titration analysis)
3-Aminobenzylamin
Formel:
C₇H₁₀N₂ Molekylvægt: 122.17 g/mol Kogepunkt: 134°C |
MDL Number:
MFCD00078355 CAS-nummer: 4403-70-7 EINECS: 412-230-2 UN nummer: 3259 ADR: 8 III |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...