2-Methoxypropen 95% stabilised
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2-Methoxy-1-propene, Isopropenyl methyl ether, 2-Methoxyprop-1-ene
Stabilization:
Stabilised with potassium carbonate
Stabilized with ca 0.5% potassium carbonate
Purity: 95%
CAS No.: 116-11-0
Molecular Formula: C4H8O
Formula Weight: 72.11
Storage Temperature: 2°C to 8°C
Flash Point: -29°(-20°F)
Refractive Index: 1.3835
Odour: Ether-like
Boiling Point: 35-36°
Beilstein Reference: 1734635
UN No.: 1993
MDL No.: MFCD00014929
Formel:
C₄H₈O Kogepunkt: 35…36 °C Massefylde: 0,758 Flammepunkt: –29 °C (–20 °F) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00014929 CAS-nummer: 116-11-0 EINECS: 204-125-3 UN nummer: 1993 ADR: 3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...