3,3-Diphenylpropylamin 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 3,3-Diphenylpropan-1-amine

B22953.14 B22953.22
B22953.14EA 1080 DKK
B22953.14 B22953.22
3,3-Diphenylpropylamin 97%
3,3-Diphenylpropylamin

Purity: 97% CAS No.: 5586-73-2 Molecular Formula: (C6H5)2CHCH2CH2NH2 Formula Weight: 211.31 Flash Point: 110°(230°F) Refractive Index: 1.5830 Melting Point: 24-29° Boiling Point: 166°/2mm Beilstein Reference: 1958595 MDL No.: MFCD00008202

Formel: C₁₅H₁₇N
Molekylvægt: 211.31 g/mol
Kogepunkt: 166 °C (2 mmHg)
Smeltepunkt: 24…29 °C
Flammepunkt: >110 °C (230 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008202
CAS-nummer: 5586-73-2
EINECS: 226-984-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR