tert-Butylchlordiphenylsilan ≥97%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 97%
CAS No.: 58479-61-1
Molecular Formula: C16H19ClSi
Formula Weight: 274.87
Flash Point: 112°(233°F)
Physical Form: Liquid
Density: 1.072
Refractive Index: 1.5680
Sensitivity: Moisture Sensitive
Odour: Pungent
Boiling Point: 90-92°/0.01mm
Beilstein Reference: 644023
UN No.: 2987
MDL No.: MFCD00000497
Formel:
C₁₆H₁₉ClSi Kogepunkt: 90…92 °C (0,01 mmHg) Massefylde: 1,072 Flammepunkt: 112 °C (233 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00000497 CAS-nummer: 58479-61-1 EINECS: 261-282-0 UN nummer: 2987 ADR: 8,II |