2-Chlorethylammoniumchlorid 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Chloroethylamine hydrochloride, 2-Chloroethylamine HCl, 2-Chloroethanamine hydrochloride

422861000 422860010 422860250
ACRO422861000EA 401 DKK
ACRO422861000 ACRO422860010 ACRO422860250
2-Chlorethylammoniumchlorid 98%
2-Chlorethylammoniumchlorid
Formel: C₂H₇Cl₂N
Molekylvægt: 115.99 g/mol
Smeltepunkt: 140…150 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012887
CAS-nummer: 870-24-6
EINECS: 212-793-2

Bestil nu

Specifikationer Testresultater

Appearance White to light beige Crystalline powder, crystals or chunks
Infrared spectrum Conforms
Melting point 140 to 150 °C
Loss on drying ≤1 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR