3,4-Dimethylbenzaldehyd 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

346060050 346060250
ACRO346060050EA 688 DKK
ACRO346060050 ACRO346060250
3,4-Dimethylbenzaldehyd 97%
3,4-Dimethylbenzaldehyde
Formel: C₉H₁₀O
Kogepunkt: 68 °C (0,8 mmHg)
Massefylde: 1,012 g/cm³
MDL Number: MFCD00016612
CAS-nummer: 5973-71-7
EINECS: 227-770-2

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5490 to 1.5510 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR