2,5-Dimethylbenzaldehyd ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Advarsel
|
Purity: 98%
CAS No.: 5779-94-2
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 87°(188°F)
Refractive Index: 1.5440
Odour: Almond-like
Boiling Point: 104-106°/14mm
Beilstein Reference: 1927303
MDL No.: MFCD00003342
Formel:
C₉H₁₀O Kogepunkt: 104…106 °C (14 mmHg) Massefylde: 0,950 Flammepunkt: 87 °C (188 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003342 CAS-nummer: 5779-94-2 EINECS: 227-303-2 |