Stopcocks for separating funnels, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286426209 286426809
SCOT286426209EA 3430 DKK
SCOT286426209 SCOT286426809
Stopcocks for separating funnels, DURAN®
Stophaner
DURAN®, borosilicate glass 3.3, clear, grounded, without key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.