Silikoneslanger, HelixMark®
Leverandør: Freudenberg Medical Europe GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60-795-22 60-795-36 60-795-05 60-795-30 60-795-21
228-0849EA 1700 DKK
228-0849 228-0850 HELI60-795-05 HELI60-795-30 HELI60-795-21
Silikoneslanger, HelixMark®
Slanger Overførselsslange
Silicone, translucent, platinum-cured.
  • Can be sterilised by autoclaving or ethylene oxide as well as by gamma radiation within the user's validation cycles and exposure levels
  • Manufactured according to GMP in an FDA-registered production facility for medical products
  • Compliant with the biocompatibility requirements of USP, Class VI
Biomedical/pharmaceutical grade tubing, manufactured by Nusil platinum-cured material. The special manufacturing process minimises flow degradation over extended periods.
Hardness (Shore durometer A): 50
Density: 1.14
Leveringsinformation: Tubing is double shrink-wrapped in crush resistant boxes. Material certification and lot traceability are included.