Tribenzylamin for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
M-Clarity™ quality level = MQ200
8.22163.0250
8.22163.9010
8.22163.0250EA
818
DKK
8.22163.0250
8.22163.9010
Tribenzylamin for synthesis, Sigma-Aldrich®
Tribenzylamin
Formel:
N(CH₂C₆H₅)₃ Molekylvægt: 287.4 g/mol Kogepunkt: 380 Smeltepunkt: 92 Massefylde: 0.9912 g/cm³ (95 °C) Flammepunkt: 65 °C (closed cup) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004773 CAS-nummer: 620-40-6 EINECS: 210-638-3 UN nummer: 1325 ADR: 4.1 III |
|
Specifikationer Testresultater
Solubility | (20 °C) insoluble |
Melting Point | 91 - 93 °C |
Molar Mass | 287.4 g/mol |
Boiling Point | 380 - 390 °C |
pH | 6.5 - 9.0 (10 g/l, H2O, 20 °C) suspension |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...