Blanks for separating and dropping funnels, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
210142404 210143606 210144405 210145401
SCOT210142404EA 1380 DKK
SCOT210142404 SCOT210143606 SCOT210144405 SCOT210145401
Blanks for separating and dropping funnels, DURAN®
Tragte Dråbetragte
DURAN®, borosilicate glass 3.3, clear.
  • cylindrical with stopper bed for ISO 4800
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For glass manipulators.