Stopcocks for burette, straight, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286408002 286408208
SCOT286408002EA 8890 DKK
SCOT286408002 SCOT286408208
Stopcocks for burette, straight, DURAN®
Stophaner
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE key.
  • Very good chemical resistance
  • Very good temperature resistance
For glass manipulators.