Straight valves 'GU', DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286058709 286058006 286058109 286058203 286058606 286058503
SCOT286058709EA 256 DKK
SCOT286058709 SCOT286058006 SCOT286058109 SCOT286058203 SCOT286058606 SCOT286058503
Straight valves 'GU', DURAN®
Stophaner
DURAN®, borosilicate glass 3.3, clear, with PTFE spindle.
  • Vacuum-tight
  • Precise dosing capacity due to fine thread
  • Safe due to unscrewing protective device
Valves are suitable for high vacuum applications, adjustable and have heavy wall side arms.