Tubing connectors, L-shaped, LaboPlast®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8707-0004 8707-0006 8707-0008 8707-0010 8707-0012 8707-0014 8713-0008 8713-0004 8713-0006 8713-0010 8713-0012 8713-0014
BURK8707-0004EA 85.5 DKK
BURK8707-0004 BURK8707-0006 BURK8707-0008 BURK8707-0010 BURK8707-0012 BURK8707-0014 229-0206 229-0204 229-0205 229-0207 229-0208 229-0209
Tubing connectors, L-shaped, LaboPlast®
Fittings og stik
PP or PVDF, translucent.
  • PP, temperature resistant up to +135 °C max.
  • PVDF, temperature resistant up to +160 °C max., highly resistant to chemicals
  • Sterilisable
Cylindrical angle hose to connect hoses. PVDF connectors with food industry approval, autoclavable and UV-resistant.