Stopcocks, single way, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286458607 286458804 286459209 286459406 286459603 286459809
SCOT286458607EA 14420 DKK
SCOT286458607 SCOT286458804 SCOT286459209 SCOT286459406 SCOT286459603 SCOT286459809
Stopcocks, single way, DURAN®
Stophaner
DURAN®, borosilicate glass 3.3, clear, with hollow glass key and glass handle.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.