Surgical gloves, latex, sterile, Gammex® Latex Sensitive
Leverandør: ANSELL MEDICAL PRODUCTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
330051075 330051055 330051060 330051095 330051085 330051070 330051090 330051080 330051065
ANSM330051075EA 857 DKK
ANSM330051075 ANSM330051055 ANSM330051060 ANSM330051095 ANSM330051085 ANSM330051070 ANSM330051090 ANSM330051080 ANSM330051065
Surgical gloves, latex, sterile, Gammex® Latex Sensitive
Handsker
These disposable gloves are specifically designed to meet the demanding requirements of microsurgery. The gloves feel like a second skin. The thin layer of latex film (20% thinner than a standard powder-free latex surgical glove) in combination with their mild textured surface provide perfect levels of sensitivity and tactility, thus ensuring a confident grip on instruments at all times.
  • Powder-free, micro-textured finish
  • Gloves feel like a second skin
  • Adhesive band at the top of the cuff maintains good contact with the surgical gown preventing the glove from rolling down
  • Brown colour reduces the adverse effect of reflections from strong light sources used during microsurgical procedures
  • Chlorination treatment inside the glove makes it easy to don in dry conditions
Recommended for all surgeries, especially delicate procedures in ophthalmology, as well as cardiovascular surgeries.
Colour: Light brown
Thickness: 0,17 mm (finger)
EN 455-1; AQL 0,65
Certificeringer: EN 455 (1-4), EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS, EN 16523-1; AQL 0,65