Tubing clamps, stop-it
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8619-0510 8619-0515 8619-0520 8619-0530
229-0117EA 64.8 DKK
229-0117 229-0118 229-0119 229-0120
Tubing clamps, stop-it
Klemmer Rørklemmer
Galvanised steel and aluminium. Used to regulate or stop the flow in tubing with liquids or gases.
  • Stepless flow regulation
  • Heat resistant
  • For hose diameters up to max. 30 mm