Dåser, bred åbning, runde, med skruelåg
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6282-2000 6282-1000 6282-0500 6282-0100 6282-0250 6282-0750 6282-1500
BURK6282-2000EA 140 DKK
BURK6282-2000 BURK6282-1000 BURK6282-0500 BURK6282-0100 BURK6282-0250 BURK6282-0750 BURK6282-1500
Dåser, bred åbning, runde, med skruelåg
Dåser Dåser, allround
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Screw lid made of PP with seal made of PE
Wide opening makes it ideally suited for storing or shipping powders and granules.