Mikrocentrifugerør, manipulationssikrede, Micrewtube®
Leverandør: Simport Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T341-2TPRTP T341-2TTP T341-4TPRTP T341-4TTP T341-5TPRTP T341-5TTP T341-6TPRTP T341-6TTP T341-7TPRTP T341-7TTP
SIMPT341-2TPRTPEA 1540 DKK
SIMPT341-2TPRTP SIMPT341-2TTP SIMPT341-4TPRTP SIMPT341-4TTP SIMPT341-5TPRTP SIMPT341-5TTP SIMPT341-6TPRTP SIMPT341-6TTP SIMPT341-7TPRTP SIMPT341-7TTP
Mikrocentrifugerør, manipulationssikrede, Micrewtube®
Rør Mikrocentrifugerør
Tubes made of PP, translucent, plain.
  • Can be used at extreme temperatures from −196 to +121 °C
  • Very good chemical resistance
  • Max. RCF: Non skirted tubes can be centrifuged up to 20000×g
Theses tubes are specially made to be used with tamperproof caps.
Ø×H: 11×44 mm.