Sample vials in Vial File®, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
224952 224891 224892 W224693 W224694 W224695 W224696 W224896 W224897 W224954 W224955
548-2161EA 1560 DKK
548-2161 548-2190 548-2191 WHEAW224693 WHEAW224694 WHEAW224695 WHEAW224696 215-3494 WHEAW224897 WHEAW224954 WHEAW224955
Sample vials in Vial File®, WHEATON®
Hætteglas
Clear vials are manufactured from low extractable borosilicate glass, amber vials made from borosilicate glass.
  • PS case with partitions, foam inserts and alphanumerical indexing card organize and protect vials
  • Clear vials conform to ASTM E438 Type I, Class A and USP Type I requirements
  • Amber vials conform to USP Type I requirements for light transmission to protect light-sensitive products
  • Caps attached to vials