4-Iodtoluen 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1-Iodo-4-methylbenzene

133940100 133940500 133941000
ACRO133940100EA 340 DKK
ACRO133940100 ACRO133940500 ACRO133941000
4-Iodtoluen 98%
4-Iodtoluen
Formel: C₇H₇I
Molekylvægt: 218.04 g/mol
Kogepunkt: 211.5
Smeltepunkt: 33…36 °C
Flammepunkt: 90 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001059
CAS-nummer: 624-31-7
EINECS: 210-841-7

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Low melting crystalline mass
Infrared spectrum Conforms
Melting point 33 to 36 °C
GC ≥97.5 %
Elemental Analysis 7.55 to 8.35 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR