3-Nitro-α,α,α,4-tetrafluortoluen 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Fluoro-3-Nitrobenzotrifluoride
200610050
200611000
ACRO200610050EA
168
DKK
ACRO200610050
ACRO200611000
3-Nitro-α,α,α,4-tetrafluortoluen 96%
3-Nitro-α,α,α,4-tetrafluortoluen
Formel:
C₇H₃F₄NO₂ Kogepunkt: 92 °C (15 mmHg) Massefylde: 1,49 g/cm³ Flammepunkt: 33 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007059 CAS-nummer: 367-86-2 EINECS: 206-702-5 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4630 (20°C, 589 nm) |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...