α-Chlor-3-nitrotoluen ≥98%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: 1-(Chloromethyl)-3-nitrobenzene, 3-Nitrobenzyl chloride, α-Chloro-m-nitrotoluene, alpha-Chloro-3-nitrotoluene, m-Nitrobenzyl chloride
A16775.09 A16775.18 A16775.30
A16775.09EA 730 DKK
A16775.09 A16775.18 A16775.30
α-Chlor-3-nitrotoluen ≥98%
α-Chlor-3-nitrotoluen
Purity: 98% CAS No.: 619-23-8 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Flash Point: 110°(230°F) Melting Point: 43-47° Boiling Point: 150°/3mm Beilstein Reference: 742794 UN No.: 3261 MDL No.: MFCD00007272
Formel: C₇H₆ClNO₂
Molekylvægt: 171.58 g/mol
MDL Number: MFCD00007272
CAS-nummer: 619-23-8