4-Fluorbenzylchlorid ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Fluorobenzyl chloride, p-Fluorobenzyl chloride, 1-Chloromethyl-4-fluorobenzene, α-Chloro-4-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene

A14686.14 A14686.22 A14686.36
A14686.14EA 431 DKK
A14686.14 A14686.22 A14686.36
4-Fluorbenzylchlorid ≥99%
4-Fluorbenzylchlorid

Purity: 99% CAS No.: 352-11-4 Molecular Formula: C7H6ClF Formula Weight: 144.58 Flash Point: 69°(156°F) Refractive Index: 1.5130 Odour: Pungent Melting Point: -18° Boiling Point: 185° Beilstein Reference: 742272 UN No.: 3265 MDL No.: MFCD00000913

Formel: C₇H₆ClF
Kogepunkt: 185 °C
Smeltepunkt: –18 °C
Massefylde: 1,214
Flammepunkt: 69 °C (156 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000913
CAS-nummer: 352-11-4
EINECS: 206-516-4
UN nummer: 3265
ADR: 8,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR